MilliporeSigma
  • Inhibition of LncRNA MALAT1 Attenuates Cerebral Ischemic Reperfusion Injury via Regulating AQP4 Expression.
Inhibition of LncRNA MALAT1 Attenuates Cerebral Ischemic Reperfusion Injury via Regulating AQP4 Expression.
European neurology (2020-11-02)
Jing Jin, Hongwei Wang, Xiaoxiao Zheng, Shangzhi Xie, Li Zheng, Renya Zhan
ABSTRACT
Stroke is one of the leading causes of mortality and disability worldwide. Long noncoding RNAs (lncRNAs) including MALAT1 have been shown to have critical roles in cerebral ischemia reperfusion injury (CIRI). However, the underlying mechanism of MALAT1 in CIRI has not been elucidated. The present study aimed to investigate the function and potential regulatory mechanism of MALAT1 in cerebral ischemic reperfusion injury. We established the middle cerebral artery occlusion (MCAO) model and oxygen-glucose deprivation/reoxygenation (OGD/RX) model in vivo and in vitro, and then Cell Counting Kit-8 (CCK-8), RT-qPCR, flow cytometry analysis, lactate dehydrogenase (LDH) analysis, and 2,3,5-triphenyltetrazolium chloride (TTC) staining were used to examine cell viability, MALAT1, aquaporin-4 (AQP4) expression, LDH release, and infarct volume, respectively. The level of AQP4 was remarkably upregulated in CIRI 24 h/48 h or OGD/RX 24 h/48 h compared with the sham group. Knockdown of AQP4 could alleviate OGD/RX-induced injury through enhancing cell viability and reducing LDH release and the rate of apoptotic cells. Furthermore, we found that MALAT1 was also increased in OGD/RX 24 h/48 h and silencing of MALAT1 could decrease AQP4. Inhibition of MALAT1 could also protect OGD/RX-induced injury, while the protective effect of MALAT1 siRNA on cerebral ischemic reperfusion was disappeared after transfection with AQP4 plasmid, indicating that MALAT1 may play a protective role in brain stroke through regulating AQP4. Taken together, our study provides evidence that MALAT1 is involved in ischemic stroke by inhibiting AQP4. Therefore, MALAT1 may serve as a potential target for therapeutic intervention in ischemic brain injury.
Sigma-Aldrich
MISSION® esiRNA, targeting human CHN1