Skip to Content
MilliporeSigma
All Photos(1)

Documents

E7284

Sigma-Aldrich

EPHA2 (561-end), active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

ECK

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

74-102 nmol/min·mg

mol wt

~72 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... EPHA2(1969)

General description

EPHA2 (ephrin receptor A2) belongs to the Eph family of receptor tyrosine kinases. It is a type I transmembrane protein, and its exoplasmic domain consists of a ligand binding domain in its N-terminal, a cysteine-rich domain, and two fibronectin type III repeats. Its cytoplasmic domain contains a juxtamembrane region, a tyrosine kinase domain, a SAM (sterile-α-motif) motif, and a PDZ-binding domain.

Biochem/physiol Actions

EPHA2 binds to the ephrin-A ligand and has diverse cellular function. EPHA2 has been shown to be an oncoprotein of importance in a range of cancers. EPHA2 is overexpressed in several human cancer types and promotes malignancy through a mechanism involving RhoA-dependent destabilization of adherens junctions. EPHA2 overexpression induces a FAK-dependent increase in MMP-2 expression and invasiveness and this process can be reversed by ligation of EPHA2.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.25 mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mark S Duxbury et al.
Biochemical and biophysical research communications, 320(4), 1096-1102 (2004-07-14)
The Eph tyrosine kinases interact with ligands of the Ephrin family and have diverse cellular functions. EphA2 has been recognized to be an oncoprotein of importance in a range of cancers. Here, we examine the effect of EphA2 overexpression and
Wei Bin Fang et al.
Journal of cell science, 121(Pt 3), 358-368 (2008-01-17)
EPHA2 receptor tyrosine kinase is overexpressed in several human cancer types and promotes malignancy. However, the mechanisms by which EPHA2 promotes tumor progression are not completely understood. Here we report that overexpression of a wild-type EPHA2, but not a signaling-defective
Linda M Reis et al.
Molecular vision, 20, 836-842 (2014-06-19)
Congenital cataracts occur in 3-4 per 10,000 live births and account for 5% to 20% of pediatric blindness worldwide. With more than 37 genes known to be associated with isolated congenital cataract, whole exome sequencing (WES) was recently introduced as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service