MilliporeSigma
  • Ligation of EphA2 by Ephrin A1-Fc inhibits pancreatic adenocarcinoma cellular invasiveness.
Ligation of EphA2 by Ephrin A1-Fc inhibits pancreatic adenocarcinoma cellular invasiveness.
Biochemical and biophysical research communications (2004-07-14)
Mark S Duxbury, Hiromichi Ito, Michael J Zinner, Stanley W Ashley, Edward E Whang
ABSTRACT
The Eph tyrosine kinases interact with ligands of the Ephrin family and have diverse cellular functions. EphA2 has been recognized to be an oncoprotein of importance in a range of cancers. Here, we examine the effect of EphA2 overexpression and ligation by chimeric Ephrin A1-Fc on the invasive phenotype of pancreatic adenocarcinoma cells. We show that EphA2 overexpression induces a FAK-dependent increase in MMP-2 expression and invasiveness. EphA2 ligation induces proteosomal degradation of EphA2, attenuates the invasive phenotype, and decreases both FAK phosphorylation and MMP-2 expression. EphA2 appears to represent a rational therapeutic target and ligation by Ephrin A1-Fc is one strategy to modulate levels of this oncoprotein.
Sigma-Aldrich
EPHA2 (561-end), active, GST tagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution