Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D8911

Sigma-Aldrich

2,3:5,6-Di-O-isopropylidene-α-D-mannofuranose

Synonym(s):

D-Mannose diacetonide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H20O6
CAS Number:
Molecular Weight:
260.28
Beilstein:
84382
MDL number:
UNSPSC Code:
12000000
Pricing and availability is not currently available.

mp

125-126 °C (dec.) (lit.)

storage temp.

−20°C

SMILES string

CC1(C)OC[C@@H](O1)[C@H]2OC(O)[C@@H]3OC(C)(C)O[C@@H]23

InChI

1S/C12H20O6/c1-11(2)14-5-6(16-11)7-8-9(10(13)15-7)18-12(3,4)17-8/h6-10,13H,5H2,1-4H3/t6?,7-,8+,9?,10?/m1/s1

InChI key

JWWCLCNPTZHVLF-RGJLLFKYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Chiral intermediate in the synthesis of glycosidase inhibitors.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fleet, G.W.J., et al.
Tetrahedron Letters, 44, 2649-2649 (1988)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service