Skip to Content
MilliporeSigma
2,3:5,6-Di-O-isopropylidene-α-D-mannofuranose 97%

2,3:5,6-Di-O-isopropylidene-α-D-mannofuranose

2,3:5,6-Di-O-isopropylidene-α-D-mannofuranose 97%
Synonym(s):
D-Mannose diacetonide
Empirical Formula (Hill Notation):
C12H20O6
CAS Number:
Molecular Weight:
260.28

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing