Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D6307

Sigma-Aldrich

6,6′-Dithiodinicotinic acid

~85%

Synonym(s):

Bis[3-Carboxypyridine] 6,6′-disulfide, Bis[3-carboxypyridine] 6,6′-disulfide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8N2O4S2
CAS Number:
Molecular Weight:
308.33
EC Number:
MDL number:
UNSPSC Code:
12000000

Assay

~85%

mp

263-265 °C (lit.)

SMILES string

OC(=O)c1ccc(SSc2ccc(cn2)C(O)=O)nc1

InChI

1S/C12H8N2O4S2/c15-11(16)7-1-3-9(13-5-7)19-20-10-4-2-8(6-14-10)12(17)18/h1-6H,(H,15,16)(H,17,18)

InChI key

GSASOFRDSIKDSN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jian-Guo Wang et al.
Bioorganic & medicinal chemistry, 15(1), 374-380 (2006-10-20)
Acetohydroxyacid synthase (AHAS; EC 2.2.1.6) catalyzes the first common step in branched-chain amino acid biosynthesis. This enzyme is an important target for the design of environmental-benign herbicides. Based on the crystal structure of AHAS/sulfonylurea complex, we have carried out computational

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service