Skip to Content
MilliporeSigma
6,6′-Dithiodinicotinic acid technical grade, 85%

6,6′-Dithiodinicotinic acid

6,6′-Dithiodinicotinic acid technical grade, 85%
Synonym(s):
Bis[3-Carboxypyridine] 6,6′-disulfide, Bis[3-carboxypyridine] 6,6′-disulfide
Empirical Formula (Hill Notation):
C12H8N2O4S2
CAS Number:
Molecular Weight:
308.33
EC Number:

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing