Documents
CLS3859BC
Corning® 1536 well plate, with generic barcode
black polystyrene plate, sterile, lid
About This Item
UNSPSC Code:
41121800
NACRES:
NB.13
material
black polystyrene plate
polystyrene plate
sterility
sterile
feature
lid
packaging
pack of 10
case of 50
maximum volume
12.5 μL
working volume
8 μL
General description
Bar Code Details
Corning barcode labels have been quality tested for optimal readability, chemical resistance and temperature variation.
  • Resistant to commonly used laboratory reagents (100% DMSO, Methanol, Ethanol, Ethyl Acetate, Methylene Chloride, Isopropanol, Acetronitrile, Ethanol/Water/TFA, Sodium Hydroxide, Hydrochloric Acid)
  • Resistant to temperature ranges of -80ºC to +52ºC
Technical Details
  • Bar Code labeling specifications
  • 2.75" x 0.25" label size
  • Code 128
  • Code 3 of 9
  • 10 mil narrow bar element (x-dimension = 0.010")
Legal Information
Corning is a registered trademark of Corning, Inc.