CLS3925
Corning® 96 well plates
flat bottom, black polystyrene, high binding surface, bag of 25, non-sterile, lid: no
Synonym(s):
96 microtiter plates, 96 multiwell plates, 96 well microplates, 96 well plates
Select a Size
$827.00
Select a Size
About This Item
$827.00
Recommended Products
material
black bottom
black polystyrene
flat bottom
polystyrene
sterility
non-sterile
feature
lid: no
skirt
plate format: 96 well standard
packaging
case of 100
bag of 25
pack of 25
manufacturer/tradename
Corning 3925
size
96 wells
well volume
360 μL
well working volume
75-200 μL
binding type
high binding surface
General description
- Black microplates have low background fluorescence and minimize light scattering
- Designed to reduce well-to-well crosstalk
- High binding surface is capable of binding medium (>10kD) and large biomolecules that possess ionic groups and/or hydrophobic regions
- Binding capacity is approximately 400 to 500ng IgG/cm2
- Ideal for immunoassays
- Without lids, lids supplied separately
- Total volume is 360μL; recommended working volume of 75 to 200μL
- Conforms to standard microplate footprint and dimensions
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)