Documents
CLS3829
Corning® multicoated microplates
well size 1536, black polystyrene (low base), flat bottom wells clear, non-sterile, lid: no
Synonym(s):
3829, multicoated
About This Item
UNSPSC Code:
41122102
NACRES:
NB.13
material
black polystyrene (low base)
flat bottom wells clear
square wells
sterility
non-sterile
feature
lid: no
manufacturer/tradename
Corning 3829
well size
1536
well volume
12.5 μL
well working volume
up to 8 μL
General description
  • No logo or lettering for improved heat sealing and ease of liquid handling
  • Low base designed for optimal bottom reader sensitivity
  • Excellent signal dynamic range due to low background fluorescence and enhanced signal intensity
  • Superior flatness minimizes variability within a plate and between plates
  • Flat bottom wells are compatible with automated dispensers
Legal Information
Corning is a registered trademark of Corning, Inc.