CLS353086
Corning® Falcon® Cell Scraper
blade L 1.8 cm, handle L 25 cm, sterile
Synonym(s):
lab equipment accessory
Select a Size
$431.00
Select a Size
About This Item
$431.00
Recommended Products
material
TPE
sterility
sterile
manufacturer/tradename
Corning 353086
technique(s)
cryopreservation: suitable
blade L
1.8 cm
handle L
25 cm
Looking for similar products? Visit Product Comparison Guide
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service