Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CLS3010

Corning® cell scrapers

blade L 1.8 cm, handle L 25 cm, sterile, case of 100

Synonym(s):

cell lifter, cell scraper

Sign Into View Organizational & Contract Pricing

Select a Size

10 EA
$59.70
100 EA
$426.00

$59.70


Please contact Customer Service for Availability


Select a Size

Change View
10 EA
$59.70
100 EA
$426.00

About This Item

UNSPSC Code:
41122103
NACRES:
NB.13

$59.70


Please contact Customer Service for Availability

sterility

sterile

packaging

case of 100

manufacturer/tradename

Corning 3010

blade L

1.8 cm

handle L

25 cm

Looking for similar products? Visit Product Comparison Guide

General description

Useful for the mechanical harvesting of cells
  • Blade design minimizes cell damage and ensures even contact with the growth surface
  • Designed for use in 25 and 75 cm² flasks
  • Individually wrapped
  • Sterilized by γ-radiation and certified nonpyrogenic

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Mostafa M Hamed et al.
Journal of medicinal chemistry, 60(7), 2853-2868 (2017-03-16)
The activation of the NF-κB transcription factor is a major adaptive response induced upon treatment with EGFR kinase inhibitors, leading to the emergence of resistance in nonsmall cell lung cancer and other tumor types. To suppress this survival mechanism, we
Juan José Dorantes-Aranda et al.
PloS one, 10(7), e0133549-e0133549 (2015-07-22)
Quantification of the role of reactive oxygen species, phycotoxins and fatty acids in fish toxicity by harmful marine microalgae remains inconclusive. An in vitro fish gill (from rainbow trout Oncorhynchus mykiss) assay was used to simultaneously assess the effect in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service