Key Documents
OMK (637-69)
90110510, owl monkey kidney, Not specified
About This Item
UNSPSC Code:
41106514
Pricing and availability is not currently available.
Product Name
OMK (637-69), from owl monkey kidney, 90110510, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA., Convention on the International Trade in Endangered Species of Wild Fauna and Flora (CITES)
biological source
owl monkey kidney
packaging
tube of 5 μg 90110510-DNA-5UG
pkg of vial of cells 90110510-1VL
growth mode
Adherent
karyotype
Not specified
morphology
Not specified
receptors
Not specified
Cell Line Origin
Owl Monkey Kidney
Cell Line Description
Derived from a normal female adult owl monkey. Sub-cultivation on over 100 occasions produced no evidence of latent infection. Used for propagation of non-human primate viruses, e.g., Herpesvirus aotus, Herpesvirus saimari and Herpesvirus ateles.
Culture Medium
EMEM (EBSS) (M2279) + 2 mM L-Glutamine (G7513) + 1% NEAA (M7145) + 10% FBS / FCS (F2442).
Subculture Routine
Split sub-confluent cultures (70-80%) 1:4 to 1:8, i.e., seeding at 1-3x10,000 cells/cm2 using 0.25% trypsin; 5% CO2; 37 °C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)