Skip to Content
MilliporeSigma
All Photos(1)

Documents

OKT9

Reactivity: Anti-CD71, human, 86022707

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Mouse x Mouse Hybridoma

ImmunoDon

CAF1 mouse spleen

Myeloma

P3X63Ag8

Reactivity

Anti-CD71, human

technique(s)

cell culture | mammalian: suitable

isotype

IgG1

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse P3X63Ag8 x Mouse CAF1 spleen

Cell Line Description

The antibody is directed against the transferrin receptor, CD71.

Immunogen

Human acute lymphoblastic leukaemia cells

Culture Medium

IDMEM + 2mM glutamine + 20% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service