Skip to Content
MilliporeSigma
All Photos(2)

Documents

AV46037

Sigma-Aldrich

Anti-KYNU (AB1) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Kynureninase (L-kynurenine hydrolase)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

52 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KYNU(8942)

Immunogen

Synthetic peptide directed towards the C terminal region of human KYNU

Application

Anti-KYNU (AB1) antibody produced in rabbit is suitable for western blotting at a concentration of 0.5μg/ml.

Biochem/physiol Actions

KYNU gene encodes a pyridoxal-5′-phosphate-(pyridoxal-P)-dependent enzyme, Kynureninase that catalyzes the cleavage of L-kynurenine into anthranilic acid and L-3-hydroxykynurenine into 3-hydroxyanthranilic acid. It facilitates the biosynthesis of NAD cofactors from tryptophan through the kynurenine pathway.

Sequence

Synthetic peptide located within the following region: LAHAVGNVELYLHDWGVDFACWCSYKYLNAGAGGIAGAFIHEKHAHTIKP

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Toma et al.
FEBS letters, 408(1), 5-10 (1997-05-12)
Kynureninase [E.C.3.7.1.3.] is one of the enzymes involved in the biosynthesis of NAD cofactors from tryptophan through the kynurenine pathway. By tryptic and CNBr digestion of purified rat liver kynureninase, we obtained about 28% of the amino acid sequence of
D Alberati-Giani et al.
European journal of biochemistry, 239(2), 460-468 (1996-07-15)
Kynureninase (L-kynurenine hydrolase), a pyridoxal-5'-phosphate-(pyridoxal-P)-dependent enzyme, catalyses the cleavage of L-kynurenine and L-3-hydroxykynurenine into anthranilic and 3-hydroxyanthranilic acids, respectively. In this report, we describe the isolation of a cDNA clone encoding human kynureninase. Degenerate oligonucleotides designed from the amino acid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service