Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C84 cell line human

12022902, human colon, Epithelial

Synonym(s):

C 84, C-84

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

C84 cell line human, 12022902

biological source

human colon

growth mode

adherent

morphology

Epithelial

shipped in

dry ice

storage temp.

−196°C

Looking for similar products? Visit Product Comparison Guide

Cell Line Description

The C84 cell line was established from a 67-year old male patient with a poorly differentiated adenocarcinoma of the caecum classified as Dukes′ stage C.

DNA Profile

STR-PCR Data:

Amelogenin: X
CSF1PO: 11,12
D13S317: 11,12
D16S539: 13
D5S818: 12
D7S820: 11
THO1: 7,9.3
TPOX: 8,9
vWA: 14,15

Culture Medium

Iscove′s Modified Dulbecco′s Medium, + 10% Fetal Bovine Serum (FBS) + 2 mM Glutamine

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4 x 10,000 cells/cm² using 0.05% trypsin or trypsin/EDTA; 5% CO2; 37°C.

C84 cells grow very slowly; following resuscitation or subculture the cells take at least 48 hours to re-attach. Cells should be left without disturbance during this time to facilitate adhesion. Centrifugation of the cells (100g x 5 min) at resuscitation to remove DMSO improves the establishment of a viable culture. Once attached, the cells grow in discrete islands and use of trypsin or trypsin/EDTA to subculture the cells (even without knocking the flask) yields large clumps. Further disaggregation may be achieved by repeatedly pipetting the cells.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service