Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

LIM2099 cell line human

12062002, human liver (metastasis), epithelial

Synonym(s):

LIM 2099, LIM-2099

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

LIM2099 cell line human, 12062002

biological source

human liver (metastasis)

growth mode

adherent

morphology

epithelial

shipped in

dry ice

storage temp.

−196°C

Looking for similar products? Visit Product Comparison Guide

Cell Line Description

Human liver metastasis, moderately differentiated sclerosing adenocarcinoma. Cell Bank Australia reference number: CBA0164.

Culture Medium

RPMI-1640 (with 2mM L-Glutamine + 25mM HEPES) + 10% FCS, Insulin 0.6 μg/ml, Hydrocortisone 1 μg/ml, 1-Thioglycerol 10μM

Subculture Routine

Optimal split ratio 1:4-1:6 (seeding density 0.8 x 104 cells/cm2). Cells incubated at 37°C with 5% CO2. Harvest the cells using 0.05% Trypsin/EDTA at 37°C for 5-10 min.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments.  Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service