Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

328/14

8062541, mouse nerve (fibroblast), Neuronal/fibroblastic

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

328/14, 08062541

biological source

mouse nerve (fibroblast)

growth mode

Adherent

karyotype

Modal No. varied with passage see reference

morphology

Neuronal/fibroblastic

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse neuroblastoma x mouse L cell fibroblast hybrid

Cell Line Description

Hybrid generated by fusion of 6-thioguanine-resistant mutant of mouse neuroblastoma N4TG3 (Sigma Catalogue number 08062515) and 8-bromodeoxyuridine-resistant mutant of mouse L cell fibroblast line B82 (Sigma Catalogue number 08062522) using inactivated Sendai virus. The fusion of N4TG3 x B82 generated two types of hybrid cells: neuronal resembling the neuroblastoma and epithelioid with clearly visible nucleus. 328/14 is of the neuronal type.

Application

Used as a neuronal Model

Culture Medium

DMEM + 2 mM glutamine + 10% Foetal Bovine Serum (FBS) + HAT (0.1 mM hypoxanthine, 10 μM aminopterin and 16 μM thymidine) Cells can be cultured without HAT provided they were cultured in between in HT medium for 3-4 days

Subculture Routine

Split sub-confluent cultures (70-80%) 1:4 to 1:8 i.e. seeding at 2-4x10,000 cells/cm2 using 0.05% trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service