Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N1E-115-1

8062511, mouse nerve, neuronal

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

N1E-115-1, 08062511

biological source

mouse nerve

growth mode

Adherent

karyotype

modal no. of chromosomes is 192 see reference

morphology

neuronal

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse neuroblastoma

Cell Line Description

A subclone of the cell line N1E-115 established by Amano et al. The cell line N1E-115 is an adrenergic cell line derived from mouse neuroblastoma C1300. (Mouse strain A/Jax). N1E-115 cells are reported to be slightly contaminated with with a cell type of the morphology of C6 glioma cells. It is recommended to use N1E-115-1, a subclone of N1E-115, that is devoid of the contamination.

Application

can be used as a neuronal model

Culture Medium

DMEM + 2 mM Glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.05% trypsin/EDTA; 5% CO2; 37°C. Population doubling approximately 20 hours.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service