Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

WERI

6070602, human eye, Grape like clusters of round cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

WERI, 06070602

biological source

human eye

growth mode

Semi-adherent

karyotype

Not specified

morphology

Grape like clusters of round cells

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Retinoblastoma

Cell Line Description

Human retinoblastoma cell line derived from a female Caucasian. The cells are semi-adherent, some neuronal differentiation can occur when subculturing. Tumourigenic in rabbits.

Culture Medium

Iscove′s Modified DMEM + 2mM Glutamine + 15% Foetal Bovine Serum (FBS) + 10ug/ml Insulin + 55uM 2-Mercaptoethanol

Subculture Routine

Split subconfluent cultures (70-80%) 1:2 to 1:4 i.e. seeding at 2-3 x100,000 cells/ml; 5% CO2; 37°C. Saturation density: 2 x 1000,000 cells/ml. Cells are semi-adherent. Scrape cells from surface; do not use trypsin.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service