Skip to Content
MilliporeSigma
All Photos(1)

Documents

UMR-106

90111314, rat bone, Not specified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

UMR-106, 90111314

biological source

rat bone

growth mode

Adherent

karyotype

Not specified

morphology

Not specified

products

Not specified

receptors

Cytosol receptors for the bone resorbing steroid hormone 1,25 -(OH)2 D3.

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Rat osteosarcoma

Cell Line Description

A cloned derivative of a transplantable rat osteogenic sarcoma. The cells retain many of the characteristics of the tumour including responsiveness to parathyroid hormone (PTH) and prostaglandins.

Application

Tumourigenicity and hormone studies

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:10 i.e. seeding at 1-3x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service