Skip to Content
MilliporeSigma
All Photos(1)

Documents

C8792

Sigma-Aldrich

Cellopentaose

≥80%

Synonym(s):

(β-D-Glc-[1→4])4-D-Glc, D(+)-Cellopentaose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H52O26
CAS Number:
Molecular Weight:
828.72
Beilstein:
79497
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic (organic)

Assay

≥80%

form

powder

storage temp.

−20°C

SMILES string

OC[C@@H](O)[C@@H](O[C@@H]1O[C@H](CO)[C@@H](O[C@@H]2O[C@H](CO)[C@@H](O[C@@H]3O[C@H](CO)[C@@H](O[C@@H]4O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4O)[C@H](O)[C@H]3O)[C@H](O)[C@H]2O)[C@H](O)[C@H]1O)[C@H](O)[C@@H](O)C=O

InChI

1S/C30H52O26/c31-1-7(37)13(39)23(8(38)2-32)53-28-20(46)16(42)25(10(4-34)50-28)55-30-22(48)18(44)26(12(6-36)52-30)56-29-21(47)17(43)24(11(5-35)51-29)54-27-19(45)15(41)14(40)9(3-33)49-27/h1,7-30,32-48H,2-6H2/t7-,8+,9+,10+,11+,12+,13+,14+,15-,16+,17+,18+,19+,20+,21+,22+,23+,24+,25+,26+,27-,28-,29-,30-/m0/s1

InChI key

FJCUPROCOFFUSR-YIQJLYQHSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Cellopentaose, a 5-glucose cellulodextrin derived from cellulose degradation similar to cellulobiose (2-glucose), cellulotriose (3-glucose), and cellulotetraose (4-glucose), is used by cellulosic bacteria as source of energy. Cellopentaose may be used to help identify, differentiate and characterize oligosaccharide metabolizing enzymes such as endoglucanases and β-glucosidase(s). Cellopentaose may be used in saccharification and ethanol fermentation research.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kyong-Cheol Ko et al.
Journal of microbiological methods, 94(3), 311-316 (2013-07-31)
Exocellulases play a key role in cleaving the accessible ends of cellulose molecules to release soluble glucose and cellobiose. To date, there have been no screens for exocellulase owing to assay protocol limitations, the high cost of substrates, and low
Leidy Lagos et al.
Microorganisms, 8(7) (2020-07-08)
The secretion of extracellular vesicles, EVs, is a common process in both prokaryotic and eukaryotic cells for intercellular communication, survival, and pathogenesis. Previous studies have illustrated the presence of EVs in supernatants from pure cultures of bacteria, including Gram-positive and
Dejan M Petrović et al.
Protein science : a publication of the Protein Society, 27(9), 1636-1650 (2018-07-05)
The catalytically crucial N-terminal histidine (His1) of fungal lytic polysaccharide monooxygenases (LPMOs) is post-translationally modified to carry a methylation. The functional role of this methylation remains unknown. We have carried out an in-depth functional comparison of two variants of a
Maíra N de Almeida et al.
Bioresource technology, 143, 413-422 (2013-07-04)
A novel multienzyme complex, E1C, and a free endoglucanase, E2 (GH5), from Fusarium verticillioides were purified. The E1C contained two endoglucanases (GH6 and GH10), one cellobiohydrolase (GH7) and one xylanase (GH10). Maximum activity was observed at 80 °C for both
Angel Angelov et al.
Scientific reports, 7(1), 17306-17306 (2017-12-13)
The discovery of novel and robust enzymes for the breakdown of plant biomass bears tremendous potential for the development of sustainable production processes in the rapidly evolving new bioeconomy. By functional screening of a metagenomic library from a volcano soil

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service