Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C7057

Nunc® Lab-Tek® II Chamber Slide system

wells 8, well area 0.7 cm2, sterile

Synonym(s):

cell culture chamber, chamber slides, microscope slides, microscopy slides

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41161502
NACRES:
NB.22
Product C7057 is not currently sold in your country. Contact Technical Service

material

polystyrene chamber

sterility

sterile

feature

CE marked

manufacturer/tradename

Nunc 154534

packaging

pack of 16 ×

well area

0.7 cm2

wells

8

working volume

0.2-0.5 mL

suitability

suitable for (cell culture; attachment and growth of cells)

Looking for similar products? Visit Product Comparison Guide

General description

Nunc® Lab-Tek® II Chamber Slide System

  • Removable, polystyrene medium chamber of 1, 2, 4 and 8-well configuration
  • Non-fluorescent microscope slide, glass (25 × 75 × 1.2 mm) with rounded corners
  • Biocompatible adhesive
  • Polystyrene cover
  • Inert hydrophobic well border printed on slide
  • Superfrost® printed writing area
  • Treated for excellent attachment and growth of cells
  • Slide separator included in each pack

Legal Information

Chamber Slide is a trademark of Thermo Fisher Scientific or its subsidiaries
Lab-Tek is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Superfrost is a registered trademark of Erie Scientific Co.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Victor Alves Carneiro et al.
Protein and peptide letters, 27(11), 1124-1131 (2020-04-17)
Ciprofloxacin (Cip) is the most commonly used quinolone in clinical practice; however large-scale use has favored the increase of multiresistant pathogenic microorganisms. Antimicrobial peptides (AMPs) appear to be a promising alternative in potentiating these conventional drugs. The aim of this
Angelo G Torrente et al.
The Journal of physiology, 595(12), 3847-3865 (2017-03-28)
Repolarizing currents through K+ channels are essential for proper sinoatrial node (SAN) pacemaking, but the influence of intracellular Ca2+ on repolarization in the SAN is uncertain. We identified all three isoforms of Ca2+ -activated small conductance K+ (SK) channels in
L S Mamsen et al.
Molecular human reproduction, 21(7), 571-582 (2015-04-30)
From early embryonic life, anti-Müllerian hormone (AMH) is produced by Sertoli cells and is essential for male sex differentiation. In females, AMH is produced by immature granulosa cells (GCs) but a definitive function in females is uncertain. We have assessed
Philip E D Chung et al.
Nature communications, 11(1), 1825-1825 (2020-04-15)
Pineoblastoma is a rare pediatric cancer induced by germline mutations in the tumor suppressors RB1 or DICER1. Presence of leptomeningeal metastases is indicative of poor prognosis. Here we report that inactivation of Rb plus p53 via a WAP-Cre transgene, commonly
Sebastian Bäumer et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 21(6), 1383-1394 (2015-01-16)
KRAS mutations are frequent driver mutations in multiple cancers. KRAS mutations also induce anti-EGFR antibody resistance in adenocarcinoma such as colon cancer. The aim of this study was to overcome anti-EGFR antibody resistance by coupling the antibody to KRAS-specific siRNA.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service