Z721336
FAST slide incubation chambers
dual well incubation chamber (for 2 Pad FAST Slides) 10/pk
Synonym(s):
hybridization chambers, incubation chambers, slide chambers
All Photos(2)
About This Item
Recommended Products
packaging
pack of 10
manufacturer/tradename
Maine Mfg 10486087
chamber L × W
79 mm × 25.4 mm
volume
300-400 μL
well L × W × D
21 mm × 21 mm × 4 mm
Related Categories
General description
Whatman array incubation chambers are designed for protein microarray applications on FAST Slides. The chambers provide a convenient way to carry out binding reactions on protein microarrays.
Used in conjunction with the FAST Frame or Chip Clip slide holder, the incubation chambers have a secure gasket design forming a tight, leak-proof seal with the FAST Slides. Simply remove the incubation chamber when the reaction is finished. These reusable chambers are recommended for use at room temperature and elevated incubation temperatures up to 76ºC. Disposable Chamber Covers are also available to prevent splashing and evaporation during incubations.
Used in conjunction with the FAST Frame or Chip Clip slide holder, the incubation chambers have a secure gasket design forming a tight, leak-proof seal with the FAST Slides. Simply remove the incubation chamber when the reaction is finished. These reusable chambers are recommended for use at room temperature and elevated incubation temperatures up to 76ºC. Disposable Chamber Covers are also available to prevent splashing and evaporation during incubations.
Other Notes
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.