Key Documents
BMSIPS2050PS1
Sigma-Aldrich
Benchmark Overhead Stirrer Propeller
flat blade, stainless steel
Select a Size
1 EACH
$136.00
$136.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EACH
$136.00
About This Item
UNSPSC Code:
41100000
NACRES:
NB.72
$136.00
Please contact Customer Service for Availability
material
stainless steel
packaging
pack of 1 ea
manufacturer/tradename
Benchmark Scientific IPS2050-P-S1
diam. × L
70 mm × 350 mm
General description
Stiring rods and propellers to fit Benchmark range of OS-20L and OS-40L overhead stirrers.
This one line, stainless steel propeller is useful for low viscosity, mid or high speed mixing.
Application
stirring, mixing
Choose from one of the most recent versions:
Certificates of Analysis (COA)