Key Documents
BMSIPS2050PT2
Sigma-Aldrich
Benchmark Overhead Stirrer Propeller
centrifugal, PTFE-coated
Select a Size
1 EACH
$214.00
$214.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EACH
$214.00
About This Item
UNSPSC Code:
41100000
NACRES:
NB.72
$214.00
Please contact Customer Service for Availability
material
PTFE-coated
packaging
pack of 1 ea
manufacturer/tradename
Benchmark Scientific IPS2050-P-T2
diam. × L
85 mm × 350 mm , open
General description
Perfect for use is small mouth, round flasks, this PTFE coated propeller is designed for mid to high speed mixing.
Stiring rods and propellers to fit Benchmark range of OS-20L and OS-40L overhead stirrers.
Application
stirring, mixing
Choose from one of the most recent versions:
Certificates of Analysis (COA)