Key Documents
BCS-600
ThawSTAR
Transport & Automated Thawing System, complete
About This Item
UNSPSC Code:
41103000
NACRES:
NB.22
feature
includes Automated Thawing System and CFT Transporter
manufacturer/tradename
BioCision BCS-600
General description
ThawSTAR automated thawing system addresses the last gap in the cell cryopreservation workflow: cell thawing. Replacing common methods of cell thawing such as swirling frozen vials in communal water baths and warming vials between hands - methods that are not standardized and potentially jeopardize the integrity and safety of the sample - ThawSTAR ensures rapid, consistent and reproducible thawing.
Utilizing patent-pending STAR sensing technology, ThawSTAR thawing system employs multiple detection mechanisms to determine vial temperature, phase change initiation, and thaw completion. Hands-free operation eliminates the guess work and subjectivity of determining the thawing end point. Highly reproducible thermal profiles mirror those obtained from a water bath without the variability and risk of contamination. Ideal for thawing cells or other biospecimens and accepts vials from -80°C freezers or LN2 storage. View the White Paper
ThawSTAR CFT Transporter is a portable solution for the handling and transport of frozen vials from long-term storage in vapor phase liquid nitrogen or in a -80°C freezer to downstream processing with the ThawSTAR automated cell thawing system. The transporter is comprised of an outer insulative foam holder, and an internal round CoolRack®; sample module that accommodates up to five 1.8 or 2.0 mL cryogenic vials.
Legal Information
CoolRack is a registered trademark of Corning, Inc.
ThawSTAR is a trademark of BioCision LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)