Skip to Content
MilliporeSigma
All Photos(1)

Documents

A5912

Supelco

N,N′-Bis(acryloyl)cystamine

~98% (HPLC)

Synonym(s):

2,2′-(Bisacrylamino)diethyl disulfide, BAC, Bis(2-acrylamidoethyl) disulfide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16N2O2S2
CAS Number:
Molecular Weight:
260.38
EC Number:
MDL number:
UNSPSC Code:
12352200

Assay

~98% (HPLC)

solubility

acetic acid: 50 mg/mL

storage temp.

−20°C

SMILES string

C=CC(=O)NCCSSCCNC(=O)C=C

InChI

1S/C10H16N2O2S2/c1-3-9(13)11-5-7-15-16-8-6-12-10(14)4-2/h3-4H,1-2,5-8H2,(H,11,13)(H,12,14)

InChI key

DJVKJGIZQFBFGS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

BAC is used as a reversible cross-linker for polyacrylamide gels, to simplify isolation of macromolecules separated on the gels. The disulfide linkage can be broken with a suitable reducing agent. A 1-g slice of a 4% (12:1) gel will dissolve in 5 mL of 100 mM DTT in TBE (pH 8.3) in less than 30 min.
BAC is used as a reversible cross-linker for polyacrylamide gels. The disulfide linkage can be broken with a suitable reducing agent. A 12:1 ratio of acrylamide:BAC is recommended for DNA purification. A one gram slice of a 4% gel will dissolve in 5 mL of 100 mM DTT, TBE solution (pH 8.3) in less than 30 min. The pH of 8.3 is optimum for rapid dissolution of the gel.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service