Skip to Content
MilliporeSigma
N,N′-Bis(acryloyl)cystamine BioReagent, suitable for electrophoresis

N,N′-Bis(acryloyl)cystamine

N,N′-Bis(acryloyl)cystamine BioReagent, suitable for electrophoresis
Synonym(s):
2,2′-(Bisacrylamino)diethyl disulfide, BAC, Bis(2-acrylamidoethyl) disulfide
Empirical Formula (Hill Notation):
C10H16N2O2S2
CAS Number:
Molecular Weight:
260.38
EC Number:

Products

Brand
Formula Weight
Physical Form
Product Line
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing