Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A4634

Sigma-Aldrich

2-Amino-6-chloropurine riboside

≥95%

Synonym(s):

(−)-2-Amino-6-chloropurine riboside, 6-Chloroguanine riboside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12ClN5O4
CAS Number:
Molecular Weight:
301.69
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:

Assay

≥95%

form

powder

mp

165-167 °C (dec.) (lit.)

solubility

water: 10 mg/mL, clear to very slightly hazy, colorless to faintly yellow

storage temp.

−20°C

SMILES string

Nc1nc(Cl)c2ncn([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)c2n1

InChI

1S/C10H12ClN5O4/c11-7-4-8(15-10(12)14-7)16(2-13-4)9-6(19)5(18)3(1-17)20-9/h2-3,5-6,9,17-19H,1H2,(H2,12,14,15)/t3-,5-,6-,9-/m1/s1

InChI key

TXWHPSZYRUHEGT-UUOKFMHZSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Amino-6-chloropurine riboside is used in the biosynthesis of mutagenic and prodrug nucleosides such as 2′-deoxy-2-(p-nitrophenyl)-adenosine and 6-deoxyacyclovir and of 6-arylthio analogues.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Matsuda et al.
Nucleic acids symposium series, 17(17), 141-143 (1986-01-01)
The reaction of 2-amino-6-chloropurine riboside with i-amyl nitrite in benzene in the presence of Cu2O, followed by treatment with NH3/MeOH gave 2-phenyladenosine (1). The crude sample of 1 was found to be mutagenic to bacteria (Salmonella typhimurium TA 98 and
Takayoshi Torii et al.
Nucleosides, nucleotides & nucleic acids, 25(4-6), 655-665 (2006-07-15)
A key compound, 2-amino-6-chlor-9-(2,3-dideoxy-3-fluoro-beta-D-erythro-pentofuranosyl)puine, was prepared from 2-amino-6-chloropurine riboside in 5 steps, then subjected to the nucleophilic displacement with benzenethiols to afford 6-arylthio congeners. These compounds showed a similar anti-HBV effect to that of 2',3' dideoxy-3'-fluoroguanosine.
Alicja Stachelska-Wierzchowska et al.
Molecules (Basel, Switzerland), 24(8) (2019-04-19)
Etheno-derivatives of guanine, O6-methylguanine, and isoguanine were prepared and purified using standard methods. The title compounds were examined as potential substrates of purine-nucleoside phosphorylases from various sources in the reverse (synthetic) pathway. It was found that 1,N2-etheno-guanine and 1,N6-etheno-isoguanine are
J T Kusmierek et al.
Acta chemica Scandinavica. Series B: Organic chemistry and biochemistry, 41(10), 701-707 (1987-11-01)
D.c. polarography of 2-amino-6-chloropurine in aqueous medium over a broad pH range revealed two diffusion waves, the first of which corresponds to reduction of the C(6)-Cl bond, leading to formation of 2-aminopurine in high yield. Condensation of the sodium salt

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service