MilliporeSigma
  • Preparative electrochemical reduction of 2-amino-6-chloropurine and synthesis of 6-deoxyacyclovir, a fluorescent substrate of xanthine oxidase and a prodrug of acyclovir.
Preparative electrochemical reduction of 2-amino-6-chloropurine and synthesis of 6-deoxyacyclovir, a fluorescent substrate of xanthine oxidase and a prodrug of acyclovir.
Acta chemica Scandinavica. Series B: Organic chemistry and biochemistry (1987-11-01)
J T Kusmierek, B Czochralska, N G Johansson, D Shugar
ABSTRACT
D.c. polarography of 2-amino-6-chloropurine in aqueous medium over a broad pH range revealed two diffusion waves, the first of which corresponds to reduction of the C(6)-Cl bond, leading to formation of 2-aminopurine in high yield. Condensation of the sodium salt of 2-aminopurine with (2-acetoxyethoxy)methyl chloride led to the two isomeric 9- and 7-(2-hydroxyethoxymethyl)-2-aminopurines. The 9- isomer, 6-deoxyacyclovir, a prodrug of acyclovir previously synthesized by another route, was readily converted to the latter by xanthine oxidase; the 7-isomer was not a substrate. The intense fluorescence of 6-deoxyacyclovir makes it a convenient fluorescent substrate for xanthine oxidase, although less sensitive than xanthine; it is shown that 2-aminopurine would be a very sensitive fluorescent substrate. The polarographic behaviour of the riboside of 2-amino-6-chloropurine was virtually identical with that of the parent purine, leading to a simple procedure for conversion of 2-amino-6-chloropurine nucleosides and acyclonucleosides to the corresponding 2-aminopurine congeners.
Sigma-Aldrich
2-Amino-6-chloropurine, ≥99%
Sigma-Aldrich
2-Amino-6-chloropurine, 97%