Skip to Content
MilliporeSigma
All Photos(1)

Documents

A211240

Sigma-Aldrich

DMT-2′Fluoro-dA(bz) Phosphoramidite

configured for MerMade

Synonym(s):

DMT-2′Fluoro-dA(bz) Amidite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C47H51FN7O7P
Molecular Weight:
875.92
MDL number:
UNSPSC Code:
41116105
NACRES:
NA.21

biological source

non-animal source (no BSE/TSE risk)

Quality Level

product line

Proligo Reagents

Assay

≥99% (31P-NMR)
≥99.0% (redox titration)

form

powder

impurities

≤0.3% water content (Karl Fischer)
≤0.5% P(III) Impurities 100-169ppm (31P-NMR)
≤0.5% single Impurity (redox titration)
≤3% residual Solvent content

color

white to off-white

suitability

conforms to structure for H-NMR
conforms to structure for LC-MS

compatibility

configured for MerMade

storage temp.

−20°C

SMILES string

CC(C)N(C(C)C)P(OCCC#N)O[C@H]([C@H](O1)COC(C2=CC=C(OC)C=C2)(C3=CC=C(OC)C=C3)C4=CC=CC=C4)[C@@H](F)[C@@H]1N5C6=NC=NC(NC(C7=CC=CC=C7)=O)=C6N=C5

InChI

1S/C47H51FN7O7P/c1-31(2)55(32(3)4)63(60-27-13-26-49)62-42-39(61-46(40(42)48)54-30-52-41-43(50-29-51-44(41)54)53-45(56)33-14-9-7-10-15-33)28-59-47(34-16-11-8-12-17-34,35-18-22-37(57-5)23-19-35)36-20-24-38(58-6)25-21-36/h7-12,14-25,29-32,39-40,42,46H,13,27-28H2,1-6H3,(H,50,51,53,56)/t39-,40-,42-,46-,63?/m1/s1

InChI key

VCCMVPDSLHFCBB-MSIRFHFKSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service