Skip to Content
MilliporeSigma
All Photos(1)

Documents

17202

Millipore

Phosphate buffer

suitable for microbiology, APHA, pH 7.2, NutriSelect® Plus

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171610
NACRES:
NA.85

Agency

according to GB 4789.10-2016
according to GB 4789.15-2016

Quality Level

sterility

non-sterile

form

powder

composition

monopotassium phosphate, 26.22 g/L
sodium carbonate, 7.78 g/L

manufacturer/tradename

NutriSelect® Plus

final pH

7.2±0.2 (25 °C)

application(s)

agriculture
environmental
food and beverages
sample preparation
veterinary
water monitoring

microbiology

Application

For the preparation of dilutions, blanks for the examination of waters, dairy products, foods, eating utensils and other specimens.

Preparation Note

Dissolve 34 g in 1 litre distilled water. Dispense and sterilize by autoclaving at 121°C for 15 minutes. The pH, after autoclaving and cooling to room temperature should be 7.2 ± 0.2.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M M Mahoney et al.
The European journal of neuroscience, 30(8), 1537-1543 (2009-10-09)
Diurnal and nocturnal animals differ with respect to the time of day at which the ovulatory surge in luteinizing hormone occurs. In some species this is regulated by the suprachiasmatic nucleus (SCN), the primary circadian clock, via cells that contain
Umut Gazi et al.
The Journal of biological chemistry, 286(10), 7822-7829 (2011-01-06)
The mannose receptor (MR) is an endocytic type I membrane molecule with a broad ligand specificity that is involved in both hemostasis and pathogen recognition. Membrane-anchored MR is cleaved by a metalloproteinase into functional soluble MR (sMR) composed of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service