Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A1135

Sigma-Aldrich

9-Aminoacridine hydrochloride hydrate

≥98%

Synonym(s):

Aminacrine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H10N2 · HCl
CAS Number:
Molecular Weight:
230.69
EC Number:
MDL number:
UNSPSC Code:
12352200

Assay

≥98%

SMILES string

O.Cl.Nc1c2ccccc2nc3ccccc13

replaced by

Product No.
Description
Pricing

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service