Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

83907

Sigma-Aldrich

Rifampicin

≥97.0% (HPLC)

Synonym(s):

3-(4-Methylpiperazinyliminomethyl)rifamycin SV, Rifampin, Rifamycin AMP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C43H58N4O12
CAS Number:
Molecular Weight:
822.94
Beilstein:
5723476
EC Number:
MDL number:
UNSPSC Code:
51101500
PubChem Substance ID:

Assay

≥97.0% (HPLC)

form

powder

color

faintly red to very dark red

solubility

chloroform: 0.25 g/5mL

antibiotic activity spectrum

mycobacteria

Mode of action

protein synthesis | interferes

storage temp.

2-8°C

SMILES string

CO[C@H]1\C=C\O[C@@]2(C)Oc3c(C)c(O)c4c(O)c(NC(=O)C(C)=C\C=C\[C@H](C)[C@H](O)[C@@H](C)[C@@H](O)[C@@H](C)[C@H](OC(C)=O)[C@@H]1C)c(\C=N\N5CCN(C)CC5)c(O)c4c3C2=O

InChI

1S/C43H58N4O12/c1-21-12-11-13-22(2)42(55)45-33-28(20-44-47-17-15-46(9)16-18-47)37(52)30-31(38(33)53)36(51)26(6)40-32(30)41(54)43(8,59-40)57-19-14-29(56-10)23(3)39(58-27(7)48)25(5)35(50)24(4)34(21)49/h11-14,19-21,23-25,29,34-35,39,49-53H,15-18H2,1-10H3,(H,45,55)/b12-11+,19-14+,22-13-,44-20+/t21-,23+,24+,25+,29-,34-,35+,39+,43-/m0/s1

InChI key

JQXXHWHPUNPDRT-WLSIYKJHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: macrolide

Application

Rifampicin is used as a selective cytochrome P4503A4 inducer. Studies have shown that rifampicin inhibits African Swine Fever Virus replication in tissue culture and inhibits the induction of tyrosine aminotransferase by cortisol in rat hepatoma cells grown in culture .

Biochem/physiol Actions

Inhibits the assembly of DNA and protein into mature virus particles.
Mode of Action: Inhibits initiation of RNA synthesis by binding to β-subunit of RNA polymerase.

Other Notes

Keep container tightly closed in a dry and well-ventilated place. Product is sensitive to light and moisture. Store under inert gas.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Lact. - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A H Dardiri et al.
Infection and immunity, 4(1), 34-36 (1971-07-01)
Vaccinia virus and African swine fever virus are deoxyribonucleic acid viruses of cytoplasmic origin. The fact that rifampin inhibits the replication of the former virus led to an investigation of its effect on African swine fever virus. The virus used
Post-transcriptional inhibition of protein synthesis by rifampicin in rat hepatoma cells.
Albert Grossman and Amal Boctor
Life Sciences, 12, 289-295 (1973)
Sandra V Kik et al.
The Journal of infectious diseases, 211 Suppl 2, S58-S66 (2015-03-15)
The potential available market (PAM) for new diagnostics for tuberculosis that meet the specifications of the high-priority target product profiles (TPPs) is currently unknown. We estimated the PAM in 2020 in 4 high-burden countries (South Africa, Brazil, China, and India)
Samy Figueiredo et al.
Antimicrobial agents and chemotherapy, 53(6), 2657-2659 (2009-03-25)
Two clonally related Acinetobacter baumannii isolates, A1 and A2, were obtained from the same patient. Isolate A2, selected after an imipenem-containing treatment, showed reduced susceptibility to carbapenems. This resistance pattern was related to insertion of the ISAba1 element upstream of
Rodney Dawson et al.
Lancet (London, England), 385(9979), 1738-1747 (2015-03-22)
New antituberculosis regimens are urgently needed to shorten tuberculosis treatment. Following on from favourable assessment in a 2 week study, we investigated a novel regimen for efficacy and safety in drug-susceptible and multidrug-resistant (MDR) tuberculosis during the first 8 weeks

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service