Skip to Content
MilliporeSigma
All Photos(1)

Documents

557303

Sigma-Aldrich

Rifampicin

≥97% (HPLC), powder (or crystals), RNA polymerase inhibitor, Calbiochem®

Synonym(s):

Rifampicin, Rifampin, 3-(4-Methylpiperazinyliminomethyl)rifamycin SV

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C43H58N4O12
CAS Number:
Molecular Weight:
822.94
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

product name

Rifampicin, Antibiotic that specifically inhibits DNA-dependent RNA polymerase in bacteria by forming an inactive complex.

Quality Level

Assay

~97-102% (by Assay)

form

powder (or crystals)

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

red-brown

shipped in

ambient

storage temp.

−20°C

InChI

1S/C43H58N4O12/c1-21-12-11-13-22(2)42(55)45-33-28(20-44-47-17-15-46(9)16-18-47)37(52)30-31(38(33)53)36(51)26(6)40-32(30)41(54)43(8,59-40)57-19-14-29(56-10)23(3)39(58-27(7)48)25(5)35(50)24(4)34(21)49/h11-14,19-21,23-25,29,34-35,39,49-53H,15-18H2,1-10H3,(H,45,55)/b12-11+,19-14+,22-13-,44-20+/t21-,23+,24+,25+,29-,34-,35+,39+,43-/m0/s1

InChI key

JQXXHWHPUNPDRT-WLSIYKJHSA-N

General description

Antibiotic that specifically inhibits DNA-dependent RNA polymerase in bacteria by forming an inactive complex. Does not affect mammalian RNA polymerase. Inhibits transcription by preventing the initial transcription complex from entering the elongation mode.
Antibiotic that specifically inhibits DNA-dependent bacterial RNA polymerase by forming an inactive complex with RNA polymerase. Does not affect mammalian RNA polymerase. Inhibits transcription by preventing the initial transcription complex from entering the elongation mode.

Biochem/physiol Actions

Primary Target
DNA-dependent RNA polymerase in bacteria

Warning

Toxicity: Harmful & Carcinogenic / Teratogenic (E)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). DMSO stock solutions are stable for up to 2 months at -20°C.

Other Notes

Jin, D.J. and Turnbough, Jr., C.L. 1994. J. Mol. Biol. 236, 72.
Levin, M.E. and Hatfull, G.F. 1993. Mol. Microbiol. 8, 277.
Wehrli, W., et al. 1968. Proc. Natl. Acad. Sci. USA61, 667.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Morten Kals et al.
Journal of the Royal Society, Interface, 21(212), 20230730-20230730 (2024-03-27)
We describe a phenotypic antibiotic susceptibility testing (AST) method that can provide an eightfold speed-up in turnaround time compared with the current clinical standard by leveraging advances in microscopy and single-cell imaging. A newly developed growth plate containing 96 agarose
Minqin Xu et al.
Journal of biomedical research, 37(6), 431-447 (2023-11-22)
cis-Diamminedichloroplatinum (CDDP) is widely used for the treatment of various solid cancers. Here we reported that CDDP increased the expression and enzymatic activities of carboxylesterase 1 (CES1) and carboxylesterase 2 (CES2), along with the upregulation of pregnane X receptor (PXR)
Lama Shamseddine et al.
iScience, 26(9), 107563-107563 (2023-09-04)
In a scenario where the discovery of new molecules to fight antibiotic resistance is a public health concern, ribosomally synthesized and post-translationally modified peptides constitute a promising alternative. In this context, the Gram-positive human gut symbiont Ruminococcus gnavus E1 produces

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service