Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

71429

Sigma-Aldrich

Sodium cyanide

BioUltra, ≥97.0% (AT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaCN
CAS Number:
Molecular Weight:
49.01
Beilstein:
3587243
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

vapor density

1.7 (vs air)

vapor pressure

1 mmHg ( 817 °C)

product line

BioUltra

Assay

≥97.0% (AT)

impurities

insoluble matter, passes filter test

pH

11.0-12.0 (25 °C, 1 M in H2O)

mp

563.7 °C (lit.)

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤100 mg/kg
phosphate (PO43-): ≤50 mg/kg
sulfate (SO42-): ≤100 mg/kg
sulfide (S2-): ≤10 mg/kg
thiocyanate (SCN-): ≤100 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤50 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤50 mg/kg
K: ≤200 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

1 M in H2O

UV absorption

λ: 260 nm Amax: 0.07
λ: 280 nm Amax: 0.045

SMILES string

[Na]C#N

InChI

1S/CN.Na/c1-2;

InChI key

RTVFYQXEHKQMKO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Enzyme inhibitor e.g. adenosylmethionine decarboxylase from E. coli

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Inhalation - Acute Tox. 1 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Met. Corr. 1 - STOT RE 1

Target Organs

Thyroid

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Purification of adenosylmethionine decarboxylase from Escherichia coli W: evidence for covalently bound pyruvate.
R B Wickner et al.
The Journal of biological chemistry, 245(8), 2132-2139 (1970-04-25)
Ping Yan et al.
The American journal of forensic medicine and pathology, 33(3), 273-275 (2011-06-08)
We report a case of homicide due to intravenous mercury injection followed by meperidine and sodium cyanide injection. A 35-year-old woman was found dead in bed at home by her husband. Reportedly, she had been sick for more than 5
Arne Wick et al.
Water research, 45(12), 3638-3652 (2011-05-17)
The sludge-water distribution of a total of 41 organic micropollutants (9 phenylurea herbicides, 11 triazines, 16 biocides and 5 UV-filters) was investigated in laboratory batch experiments with fresh secondary sludge taken from a municipal WWTP. Sorption kinetics as well as
Monica Lemus et al.
Nitric oxide : biology and chemistry, 25(4), 387-395 (2011-10-11)
Previous work has shown that the carotid body glomus cells can function as glucose sensors. The activation of these chemoreceptors, and of its afferent nucleus in the brainstem (solitary tract nucleus - STn), induces rapid changes in blood glucose levels
Fei Chen et al.
Clinical toxicology (Philadelphia, Pa.), 49(2), 121-123 (2011-03-05)
Cyanide is considered one of the most lethal poisons, reports on visual changes after acute cyanide poisoning are rare due to a low rate of survival of those having committed suicides. A 30-year-old man developed visual loss in both eyes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service