MilliporeSigma
  • Homicide due to intravenous metallic mercury injection followed by sodium cyanide injection.
Homicide due to intravenous metallic mercury injection followed by sodium cyanide injection.
The American journal of forensic medicine and pathology (2011-06-08)
Ping Yan, Guangzhao Huang, Daoquan Li, Ling Li
ABSTRACT
We report a case of homicide due to intravenous mercury injection followed by meperidine and sodium cyanide injection. A 35-year-old woman was found dead in bed at home by her husband. Reportedly, she had been sick for more than 5 months. Initial death investigation revealed no evidence of foul play. Her death was believed to be natural. Therefore, her body was buried without an autopsy. Two months after death, her family requested an autopsy because they suspected her physician husband killed her. Her body was exhumed, and an autopsy was performed. Postmortem examination revealed numerous metallic mercury globules in the pulmonary arteries. Toxicological analysis revealed a high concentration of mercury in the tissue samples of the lungs, liver, heart, and kidney. In addition, cyanide and meperidine were also found in the heart and liver. The detailed case history and postmortem examination findings are described.
Sigma-Aldrich
Sodium cyanide, reagent grade, 97%
Sigma-Aldrich
Sodium cyanide, ACS reagent, ≥95.0%