Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

69336

Sigma-Aldrich

Spectral Fluorescence Standard Kit (certified by BAM)

Synonym(s):

Certified standard for the determination of fluorescence wavelengths, Determination of fluorescence wavelengths, certified standard, Emission Wavelength Standard Kit (certified, BAM)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

storage temp.

2-8°C

Quality Level

General description

This improved version of the Spectral Fluorescence Standards Kit provides a simple, flexible, and traceable calibration tool for the determination and control of the relative spectral responsivity of fluorescence instruments. The kit is optimized for spectrofluorometers and can be employed for a broad variety of measurement geometries. With proper consideration of the underlying measurement principle, it can be adapted to the calibration of other types of fluorescence instruments.

Application

Suitable for the spectral range of 300-760 nm

Components

contains:
Fluorescence Spectral Standard A
Fluorescence Spectral Standard B′
Fluorescence Spectral Standard C
Fluorescence Spectral Standard D
Fluorescence Spectral Standard E
Ethanol Spectral
BAM-Certificate
CD

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service