Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.19302

Supelco

Verification Standard

Sign Into View Organizational & Contract Pricing

Select a Size

1 UNIT
$346.00

$346.00


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
1 UNIT
$346.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

$346.00


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Move 100

storage temp.

15-25°C

General description

for Spectroquant® Move 100 Colorimeter and Spectroquant Multy Colorimeter

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameCorrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

78.8 °F

Flash Point(C)

26 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service