Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

56499C

SAFC

Dulbecco′s Modified Eagle′s Medium/High Modified

with 4500 mg/L dextrose, with 4.0 mM L-glutamine, with 110 mg/L sodium pyruvate, without sodium bicarbonate, powder, suitable for cell culture

Pharma Manufacturing

Synonym(s):

Dulbecco’s Modified Eagle’s Medium - high glucose, DME, DMEM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.71

description

for research or for further manufacturing use

Quality Level

form

powder

technique(s)

cell culture | mammalian: suitable

components

phenol red: yes
sodium pyruvate: yes
L-glutamine: yes
glucose: high

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Dulbecco′s Modified Eagle′s Medium (DMEM) is a modification of Basal Medium Eagle (BME) that contains four-fold concentrations of the amino acids and vitamins. The original formulation contained 1000 mg/L of glucose and was used to culture embryonic mouse cells. Since then, it has been modified in several ways to support primary cultures of mouse and chicken cells, as well as a variety of normal and transformed cells. Each of these media offers a different combination of L-glutamine and sodium pyruvate. Additionally, the glucose levels have been raised to 4500 mg/L, contributing to the name "DMEM/High".

Quantity

Formulated to contain 13.5 grams of powder per liter of medium.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marko Knoll et al.
Nature communications, 8(1), 2115-2115 (2017-12-14)
Brown adipose tissue (BAT) metabolism influences glucose homeostasis and metabolic health in mice and humans. Sympathetic stimulation of β-adrenergic receptors in response to cold induces proliferation, differentiation, and UCP1 expression in pre-adipocytes and mature brown adipocytes. Here we show that
Mengxi Jiang et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(26), 15055-15065 (2020-06-20)
Phosphocholine phosphatase-1 (PHOSPHO1) is a phosphocholine phosphatase that catalyzes the hydrolysis of phosphocholine (PC) to choline. Here we demonstrate that the PHOSPHO1 transcript is highly enriched in mature brown adipose tissue (BAT) and is further induced by cold and isoproterenol
Maria Masnata et al.
Acta neuropathologica, 137(6), 981-1001 (2019-02-23)
In recent years, evidence has accumulated to suggest that mutant huntingtin protein (mHTT) can spread into healthy tissue in a prion-like fashion. This theory, however, remains controversial. To fully address this concept and to understand the possible consequences of mHTT spreading

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service