Skip to Content
MilliporeSigma
All Photos(1)

Documents

50796

Sigma-Aldrich

Maltulose monohydrate

≥98.0% (HPLC)

Synonym(s):

4-O-α-D-Glucopyranosyl-D-fructose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O11 · H2O
CAS Number:
Molecular Weight:
360.31
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

Assay

≥98.0% (HPLC)

mp

137 °C (dec.) (lit.)

SMILES string

O.OC[C@H]1O[C@H](O[C@@H]2[C@H](O)CO[C@@](O)(CO)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C12H22O11.H2O/c13-1-5-6(16)7(17)8(18)11(22-5)23-9-4(15)2-21-12(20,3-14)10(9)19;/h4-11,13-20H,1-3H2;1H2/t4-,5-,6-,7+,8-,9-,10+,11-,12+;/m1./s1

InChI key

NNDCKRNDWLPICC-MDKWJHPDSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Maltulose, an α(1,4) linked disaccharide of D-glucose and D-fructose, is used as a reference in analytical procedures to determine the extent of nonenzmatic browning induced by processes such as heating. Maltulose is used to identify, differentiate and characterize α-α-glucosidase(s).

Biochem/physiol Actions

A sugar molecule found in honey

Other Notes

To gain a comprehensive understanding of our extensive range of Disaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F W Parrish et al.
Journal of bacteriology, 96(1), 227-233 (1968-07-01)
Spores of the fungus Myrothecium verrucaria are cryptic to maltose and isomaltose. Induction of a transport system can be effected by several sugars whose order of effectiveness is: turanose > maltulose > sucrose > d-arabinose, d-fructose, nigerose, maltotriulose, kestose >
Valle Morales et al.
Journal of agricultural and food chemistry, 52(22), 6732-6736 (2004-10-28)
Nonenzymic browning reactions in commercial infant formulas were evaluated through their furosine content as well as the isomeric disaccharides formed during processing. Lactulose was observed only in samples containing appreciable amounts of lactose, whereas maltulose was present in all samples
J Thompson et al.
The Journal of biological chemistry, 276(40), 37415-37425 (2001-07-27)
Klebsiella pneumoniae is presently unique among bacterial species in its ability to metabolize not only sucrose but also its five linkage-isomeric alpha-d-glucosyl-d-fructoses: trehalulose, turanose, maltulose, leucrose, and palatinose. Growth on the isomeric compounds induced a protein of molecular mass approximately

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service