Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

T0299

Sigma-Aldrich

α,β-Trehalose

≥98%

Synonym(s):

α-D-Glucopyranosyl β-D-glucopyranoside, Neotrehalose

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$842.00

$842.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
$842.00

About This Item

Empirical Formula (Hill Notation):
C12H22O11
CAS Number:
Molecular Weight:
342.30
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

$842.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

plant

Assay

≥98%

form

crystalline powder

technique(s)

HPLC: suitable

SMILES string

OC[C@H]1O[C@H](O[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C12H22O11/c13-1-3-5(15)7(17)9(19)11(21-3)23-12-10(20)8(18)6(16)4(2-14)22-12/h3-20H,1-2H2/t3-,4-,5-,6-,7+,8+,9-,10-,11-,12+/m1/s1

InChI key

HDTRYLNUVZCQOY-BTLHAWITSA-N

Other Notes

To gain a comprehensive understanding of our extensive range of Disaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service