Skip to Content
MilliporeSigma
All Photos(2)

Documents

308-05A

Sigma-Aldrich

Human Internal Thoracic Artery Endothelial Cells: HITAEC, adult

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

biological source

human intrathoratic artery (normal)

Quality Level

packaging

pkg of 500,000 cells

manufacturer/tradename

Cell Applications, Inc

growth mode

Adherent

karyotype

2n = 46

morphology

Endothelial

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

diabetes; cardiovascular diseases

shipped in

dry ice

storage temp.

−196°C

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.

HITAEC has been used to investigate oral pathogens travel through the body by infecting dendritic cells and inducing them to migrate through endothelial layers by causing upregulation of CXCR4 (Miles, 2014). To identify genes that make Human Internal Thoracic Arteries resistant to atherosclerosis, HITAEC and HCAEC (derived from the athero-prone human coronary arteries) were subjected to microarray analysis (Archaki, 2012). Twenty-nine genes were found to be expressed differently between HITAEC and HCAEC; one of them (TES) was shown to be downregulated in patients with coronary artery disease, resulting in increased monocyte adhesion, endothelial cell motility and transendothelial migration of monocytes.

Cell Line Origin

Artery

Application

coronary artery bypass grafting, myocardial revascularization, endothelial cell motility, transendothelial migration, gene expression, contractile responses, receptor stimulation, cell relaxation, supply of oxygenated blood to chest wall and breasts,

Components

Basal Medium containing 10% FBS & 10% DMSO

Preparation Note

  • 2nd passage, >500,000 cells in Basal Medium containing 10% FBS & 10% DMSO
  • Can be cultured at least 10 doublings

Subculture Routine

Please refer to the HITAEC Culture Protocol.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service