Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SCCE020

Sigma-Aldrich

Human Epidermal Keratinocytes, Neonatal

Human Neonatal Epidermal Keratinocytes provide an ideal serum-free culture model for the study of wound healing, toxicology, or biology when grown in our recommended medium.

Sign Into View Organizational & Contract Pricing

Select a Size

1 VIAL
$851.00

$851.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 VIAL
$851.00

About This Item

UNSPSC Code:
41106514
eCl@ss:
32011203
NACRES:
NA.71

$851.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

human epithelium (keratinocytes)

Quality Level

packaging

vial of 500,000 cells

manufacturer/tradename

Chemicon®
EpiGRO®
Millipore

morphology

(epidermal)

technique(s)

cell culture | mammalian: suitable

shipped in

liquid nitrogen

General description

Millipore’s Human Neonatal Epidermal Keratinocytes, when grown in our recommended medium, provide an ideal serum-free culture model for the study of wound healing, toxicology, or biology. These cells are not exposed to antimicrobials or phenol red when cultured in EpiGRO Human Keratinocyte Complete Media Kit. Human Neonatal Epidermal Keratinocytes are cryopreserved at passage 1 to ensure highest viability and plating efficiency.

This product is for Research Use Only. This product is not approved for human or veterinary use, or for use in in vitro diagnostic or clinical procedures
Product Source: Human Neonatal Keratinocyte

Cell Line Origin

Keratinocytes

Cell Line Description

Epithelial Cells

Application

Please see data sheet for detailed directions on Thawing, Plating , Feeding, Passaging, etc.
Research Category
Stem Cell Research

Packaging

500,000 cells

Components

SCCE020

Quality

Cells are negative for HIV (1,2), HBV, HCV, bacteria, mycoplasma and
fungal contamination.

Further growth: Guaranteed to provide 15 or more doublings when grown in the appropriate EpiGRO Medium.

Storage and Stability

To maintain the cells’ integrity, Millipore recommends unpacking the products immediately upon receipt and storing at a temperature lower than -150°C or in liquid nitrogen vapor phase.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
EPIGRO is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service