Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

137269

SAFC

BIS-TRIS propane

EMPROVE® EVOLVE

Pharma Manufacturing

Synonym(s):

BIS-TRIS propane, 1,3-Bis[tris(hydroxymethyl)methylamino]propane

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$724.00
3 X 100 G
$2,468.00
1372691000
$4,116.00
1372695000
$20,729.00
10 KG
$31,597.00

$724.00


Please contact Customer Service for Availability


Select a Size

Change View
100 G
$724.00
3 X 100 G
$2,468.00
1372691000
$4,116.00
1372695000
$20,729.00
10 KG
$31,597.00

About This Item

Linear Formula:
CH2[CH2NHC(CH2OH)3]2
CAS Number:
Molecular Weight:
282.33
Beilstein:
1786109
MDL number:
UNSPSC Code:
12161705

$724.00


Please contact Customer Service for Availability

biological source

synthetic

Quality Level

product line

EMPROVE® EVOLVE

form

powder

technique(s)

cell culture | mammalian: suitable

pH

6.3-9.5

useful pH range

6.3-9.5

pKa (25 °C)

(1) 6.8, (2) 9.0

pKa 

(1) 6.8, (2) 9.0

mp

164-165 °C (lit.)
164-165 °C (lit.)

suitability

suitable for manufacturing use

foreign activity

Cytotoxicity, DNase, NICKase, RNase, and Protease; tested

storage temp.

2-30°C

SMILES string

OCC(CO)(CO)NCCCNC(CO)(CO)CO

InChI

1S/C11H26N2O6/c14-4-10(5-15,6-16)12-2-1-3-13-11(7-17,8-18)9-19/h12-19H,1-9H2

InChI key

HHKZCCWKTZRCCL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Bis Tris Propane is a unique biological buffer also referred to as a “Universal” buffer, which means the buffer capacity covers a wide range. Bis Tris Propane has two pKas of 6.3 and 9.5, which expands the buffering range from 5.0-10.5-making this a versatile buffer for biological systems. Bis Tris Propane is considered to be non-toxic to culture cell lines and provides high-solution clarity.A review of DNA polymerase techniques lists bis tris propane as a suitable buffer for PCR methods as a universal buffer pH gradient is employed.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service