Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

137272

SAFC

Guanidine hydrochloride

EMPROVE® EVOLVE

Pharma Manufacturing

Synonym(s):

Guanidine hydrochloride, Aminoformamidine hydrochloride, Aminomethanamidine hydrochloride, Guanidinium chloride

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$308.00
3 X 100 G
$1,090.00
2 KG
$1,745.00
5 KG
$3,745.00
25 KG
$10,232.00
50 KG
$18,185.00

$308.00


Please contact Customer Service for Availability


Select a Size

Change View
100 G
$308.00
3 X 100 G
$1,090.00
2 KG
$1,745.00
5 KG
$3,745.00
25 KG
$10,232.00
50 KG
$18,185.00

About This Item

Linear Formula:
NH2C(=NH)NH2 · HCl
CAS Number:
Molecular Weight:
95.53
Beilstein:
3591990
MDL number:
UNSPSC Code:
12352107

$308.00


Please contact Customer Service for Availability

biological source

synthetic

Quality Level

product line

EMPROVE® EVOLVE

form

powder

impurities

Endotoxin, microbial, and trace metals; tested

pH

(25 °C, 4.6 - 6/573 g/L)

mp

180-185 °C (lit.)
180-185 °C (lit.)

solubility

water: 6 M at 20 °C, clear, colorless

density

1.3 g/cm3 (lit.)
1.3 g/cm3 (lit.)

suitability

suitable for manufacturing use

storage temp.

2-30°C

SMILES string

Cl[H].NC(N)=N

InChI

1S/CH5N3.ClH/c2-1(3)4;/h(H5,2,3,4);1H

InChI key

PJJJBBJSCAKJQF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Strong chaotropic agent useful for the denaturation and subsequent refolding of proteins. This strong denaturant can solubilize insoluble or denatured proteins such as inclusion bodies. This can be used as the first step in refolding proteins or enzymes into their active form. Urea and dithiothreitol (DTT) may also be necessary.

Application

Strong chaotropic agent useful for the denaturation and subsequent refolding of proteins. This strong denaturant can solubilize insoluble or denatured proteins such as inclusion bodies. This can be used as the first step in refolding proteins or enzymes into their active form. Urea and dithiothreitol (DTT) may also be necessary.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service