Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

01912

Sigma-Aldrich

N-Acetyl-D-sphingosine

≥98.0% (TLC)

Synonym(s):

(2S,3R,4E)-2-(Acetylamino)-4-octadecene-1,3-diol, N-Acetoyl-D-erythro-sphingosine, Acetyl ceramide, C2 Ceramide (d18:1/2:0), C2 ceramide

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$344.00
250 MG
$1,120.00

$344.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
$344.00
250 MG
$1,120.00

About This Item

Empirical Formula (Hill Notation):
C20H39NO3
Molecular Weight:
341.53
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.85

$344.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥98.0% (TLC)

lipid type

sphingolipids

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCC\C=C\[C@H](O)[C@@H](CO)NC(C)=O
CCCCCCCCCCCCC\C=C\[C@H](O)[C@@H](CO)NC(C)=O

InChI

1S/C20H39NO3/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-20(24)19(17-22)21-18(2)23/h15-16,19-20,22,24H,3-14,17H2,1-2H3,(H,21,23)/b16-15+/t19-,20+/m1/s1

InChI key

BLTCBVOJNNKFKC-HEQKZDDYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Cell-permeable, biologically active ceramide. It induces differentiation and apoptosis in cells and has been shown to activate protein phosphatases.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gokhan Kus et al.
In vitro cellular & developmental biology. Animal, 51(10), 1056-1063 (2015-07-15)
Ceramidases are key enzymes that decrease ceramide levels in cells. A reduction in ceramide concentration impairs ceramide signalling, and results in apoptosis resistance in cancer cells. This study investigates the potential for ceranib-2, a novel ceramidase inhibitor, to affect the
Wei Wang et al.
PloS one, 10(4), e0119005-e0119005 (2015-04-24)
Conjugated linoleic acid (CLA) reduces adiposity in human and mouse adipocytes. This outcome is achieved through a variety of biological responses including increased energy expenditure and fatty acid oxidation, increased inflammation, repression of fatty acid biosynthesis, attenuated glucose transport, and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service