MilliporeSigma
  • Sphingolipids are required for efficient triacylglycerol loss in conjugated linoleic Acid treated adipocytes.
Sphingolipids are required for efficient triacylglycerol loss in conjugated linoleic Acid treated adipocytes.
PloS one (2015-04-24)
Wei Wang, Michael Fromm
ABSTRACT
Conjugated linoleic acid (CLA) reduces adiposity in human and mouse adipocytes. This outcome is achieved through a variety of biological responses including increased energy expenditure and fatty acid oxidation, increased inflammation, repression of fatty acid biosynthesis, attenuated glucose transport, and apoptosis. In the current study, profiling of 261 metabolites was conducted to gain new insights into the biological pathways responding to CLA in 3T3-L1 adipocytes. Sphinganine and sphingosine levels were observed to be highly elevated in CLA treated adipocytes. Exogenous chemicals that increased endogenous ceramide levels decreased lipid levels in adipocytes, and activated AMP-activated protein kinase (AMPK) as well as NF-κB, both of which are typically activated in CLA treated adipocytes. Concurrent inhibition of ceramide de novo biosynthesis and recycling from existing sphingolipid pools attenuated the lipid lowering effect normally associated with responses to CLA, implicating ceramides as an important component of the lipid lowering response in CLA treated adipocytes.
Product Description
Supelco
Potassium hydroxide concentrate, 0.1 M KOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Dorsomorphin, ≥98% (HPLC)
Supelco
Fumonisin B1, reference material, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Bovine Calf Serum, Iron-Fortified, USA origin, from formula-fed bovine calves, sterile-filtered, suitable for cell culture
Sigma-Aldrich
N-Acetyl-D-sphingosine, ≥98.0% (TLC)
Sigma-Aldrich
Serum Triglyceride Determination Kit, 1 kit sufficient for 250 tests
Sigma-Aldrich
SKI II, ≥98% (HPLC), solid
Supelco
Fumonisin B1 solution, ~50 μg/mL in acetonitrile: water, analytical standard
Sigma-Aldrich
Insulin from bovine pancreas, γ-irradiated, BioXtra, suitable for cell culture, potency: ≥20 units/mg (USP units), lyophilized powder
Sigma-Aldrich
N-Acetyl-D-sphingosine, ≥97% (TLC), powder
Sigma-Aldrich
Myriocin from Mycelia sterilia, ≥98% (HPLC), powder
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Serine, European Pharmacopoeia (EP) Reference Standard
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Potassium hydroxide, tested according to Ph. Eur.
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
bisBenzimide H 33342 trihydrochloride, ≥98% (HPLC and TLC)
Sigma-Aldrich
DL-Serine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
DL-Serine, ≥98% (TLC)
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Fumonisin B1 from Fusarium moniliforme, ≥98% (HPLC)
Sigma-Aldrich
Deoxyribonucleic acid sodium salt from salmon testes
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Potassium hydroxide, anhydrous, ≥99.95% trace metals basis