Skip to Content
MilliporeSigma
All Photos(1)

Documents

Y0000463

Vinorelbine tartrate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Vinorelbine ditartrate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H54N4O8 · 2C4H6O6
CAS Number:
Molecular Weight:
1079.11
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

vinorelbine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−70°C

InChI

1S/C45H54N4O8.2C4H6O6/c1-8-27-19-28-22-44(40(51)55-6,36-30(25-48(23-27)24-28)29-13-10-11-14-33(29)46-36)32-20-31-34(21-35(32)54-5)47(4)38-43(31)16-18-49-17-12-15-42(9-2,37(43)49)39(57-26(3)50)45(38,53)41(52)56-7;2*5-1(3(7)8)2(6)4(9)10/h10-15,19-21,28,37-39,46,53H,8-9,16-18,22-25H2,1-7H3;2*1-2,5-6H,(H,7,8)(H,9,10)/t28?,37-,38+,39+,42+,43+,44-,45-;2*1-,2-/m011/s1

InChI key

CILBMBUYJCWATM-NPJYPKOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Vinorelbine tartrate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Carc. 2 - Muta. 2 - Repr. 1B - STOT RE 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of human cytochrome P450 isoenzymes involved in the metabolism of vinorelbine.
Beulz?Riche D, et al.
Fundamental & Clinical Pharmacology, 19(5), 545-553 (2005)
Acute respiratory failure caused by vinorelbine tartrate in a patient with non-small cell lung cancer.
Uoshima N, et al.
Internal medicine (Tokyo, Japan), 40(8), 779-782 (2001)
Development and Validation of New RPHPLC Method for the Estimation of Vinorelbine in Pharmaceutical Dosage Forms.
Gandhi B M, et al.
International Journal of Research in Pharmacy and Chemistry, 201(3), 3-3 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service